ENA321761775 Enamine

2-chloro-N-[1-(2-oxo-2,3-dihydro-1H-indol-5-yl)ethyl]acetamide

  • MDL number MFCD18483145
  • Cas Number : 1311313-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49028
Availability details for
Quantity
  • Availability
  • Add to Cart