ENA321761778 Enamine

3-ethylpyrido[3,2-d][1,2]oxazole-5-carboxylic acid

  • MDL number MFCD18838643
  • Cas Number : 1306605-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193...195
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52766

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52766
Availability details for
Quantity
  • Availability
  • Add to Cart