ENA321761781 Enamine

1-{5-[(dimethylamino)methyl]-1,2,4-oxadiazol-3-yl}cyclopentan-1-amine dihydrochloride

  • MDL number MFCD18917234
  • Cas Number : 1311317-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53839

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53839
Availability details for
Quantity
  • Availability
  • Add to Cart