ENA321761783 Enamine

5-ethyl-3-({octahydropyrrolo[3,4-c]pyrrol-2-yl}methyl)-1,2,4-oxadiazole dihydrochloride

  • MDL number MFCD18483149
  • Cas Number : 1311318-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55012
Availability details for
Quantity
  • Availability
  • Add to Cart