ENA321761787 Enamine

2,2,2-trifluoroethyl N-(1-acetylpiperidin-4-yl)carbamate

  • MDL number MFCD18483153
  • Cas Number : 1311316-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56578
Availability details for
Quantity
  • Availability
  • Add to Cart