ENA321761788 Enamine

2,2,2-trifluoroethyl N-[3-(pyrrolidin-1-yl)propyl]carbamate

  • MDL number MFCD18838646
  • Cas Number : 1306603-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56579
Availability details for
Quantity
  • Availability
  • Add to Cart