ENA321761789 Enamine

ethyl 2-(2-{[(2,2,2-trifluoroethoxy)carbonyl]amino}-1,3-thiazol-4-yl)acetate

  • MDL number MFCD18785443
  • Cas Number : 1311314-73-4
  • Molecular Weight : 312.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56588
Availability details for
Quantity
  • Availability
  • Add to Cart