ENA321761790 Enamine

2,2,2-trifluoroethyl N-[4-(pyridin-3-yl)-1,3-thiazol-2-yl]carbamate

  • MDL number MFCD18483154
  • Cas Number : 1311316-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56631
Availability details for
Quantity
  • Availability
  • Add to Cart