ENA321761791 Enamine

2,2,2-trifluoroethyl N-(4-methyl-1,3-benzothiazol-2-yl)carbamate

  • MDL number MFCD18838647
  • Cas Number : 1306604-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56641
Availability details for
Quantity
  • Availability
  • Add to Cart