ENA321761794 Enamine

2,2,2-trifluoroethyl N-(4-methoxy-1,3-benzothiazol-2-yl)carbamate

  • MDL number MFCD18483155
  • Cas Number : 1311314-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56682
Availability details for
Quantity
  • Availability
  • Add to Cart