ENA321761795 Enamine

2,2,2-trifluoroethyl N-[4-(3-fluorophenyl)-1,3-thiazol-2-yl]carbamate

  • MDL number MFCD18483156
  • Cas Number : 1311314-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-56702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56702
Availability details for
Quantity
  • Availability
  • Add to Cart