ENA321761806 Enamine

1-{5-[(dimethylamino)methyl]-1,2,4-oxadiazol-3-yl}cyclopentan-1-amine

  • MDL number MFCD18917236
  • Cas Number : 1311318-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58607
Availability details for
Quantity
  • Availability
  • Add to Cart