ENA321761812 Enamine

3-(chloromethyl)-5-(4-phenyloxan-4-yl)-1,2,4-oxadiazole

  • MDL number MFCD18917237
  • Cas Number : 1311316-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63668

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63668
Availability details for
Quantity
  • Availability
  • Add to Cart