ENA321761815 Enamine

2-(benzylsulfanyl)propanedial

  • MDL number MFCD18917238
  • Cas Number : 1311318-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63918
Availability details for
Quantity
  • Availability
  • Add to Cart