ENA321761821 Enamine

2,2,2-trifluoroethyl 3-(dimethylamino)-1-oxo-1,2-dihydroisoquinoline-4-carboxylate

  • MDL number MFCD18483167
  • Cas Number : 1311316-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70075
Availability details for
Quantity
  • Availability
  • Add to Cart