ENA321761831 Enamine

2-[(1H-pyrazol-1-yl)methyl]morpholine

  • MDL number MFCD18483168
  • Cas Number : 1311317-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71170
Availability details for
Quantity
  • Availability
  • Add to Cart