ENA321761834 Enamine

2-[but-3-yn-1-yl(methyl)amino]acetic acid hydrochloride

  • MDL number MFCD18838664
  • Cas Number : 1306606-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71532
Availability details for
Quantity
  • Availability
  • Add to Cart