ENA321761835 Enamine

4-(aminomethyl)-N-[2-(pyrrolidin-1-yl)ethyl]benzamide dihydrochloride

  • MDL number MFCD18483170
  • Cas Number : 1028338-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71647
Availability details for
Quantity
  • Availability
  • Add to Cart