ENA321761860 Enamine

3-amino-1-(5-nitropyridin-2-yl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD18483186
  • Cas Number : 1311313-99-1
  • Molecular Weight : 230.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73526
Availability details for
Quantity
  • Availability
  • Add to Cart