ENA321761862 Enamine

1-(5-methyl-1,3,4-oxadiazol-2-yl)ethan-1-ol

  • MDL number MFCD18785456
  • Cas Number : 1311316-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73548
Availability details for
Quantity
  • Availability
  • Add to Cart