ENA321761863 Enamine

[4-(2-chloroacetyl)phenyl]urea

  • MDL number MFCD18483188
  • Cas Number : 861520-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73550
Availability details for
Quantity
  • Availability
  • Add to Cart