ENA321761866 Enamine

methyl 2-{4-oxo-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-1-yl}acetate

  • MDL number MFCD18483192
  • Cas Number : 1311316-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73619
Availability details for
Quantity
  • Availability
  • Add to Cart