ENA321761867 Enamine

4-[(3-methyl-1,2,4-oxadiazol-5-yl)methoxy]aniline hydrochloride

  • MDL number MFCD18838670
  • Cas Number : 1306605-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73639

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73639
Availability details for
Quantity
  • Availability
  • Add to Cart