ENA321761879 Enamine

[1-(phenylsulfanyl)cyclopentyl]methanamine

  • MDL number MFCD18483203
  • Cas Number : 1311314-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73741
Availability details for
Quantity
  • Availability
  • Add to Cart