ENA321761880 Enamine

2,6-dichloroquinoline-4-carbonitrile

  • MDL number MFCD18483205
  • Cas Number : 50504-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73748
Availability details for
Quantity
  • Availability
  • Add to Cart