ENA321761890 Enamine

2-(ethanesulfinyl)ethan-1-amine hydrochloride

  • MDL number MFCD18483214
  • Cas Number : 176240-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73799
Availability details for
Quantity
  • Availability
  • Add to Cart