ENA321761892 Enamine

1-(5-fluoro-1-methyl-1H-1,3-benzodiazol-2-yl)methanamine dihydrochloride

  • MDL number MFCD18483215
  • Cas Number : 1311314-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73805
Availability details for
Quantity
  • Availability
  • Add to Cart