ENA321761899 Enamine

1-(aminomethyl)-4-tert-butylcyclohexan-1-ol hydrochloride

  • MDL number MFCD18483223
  • Cas Number : 91249-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73845
Availability details for
Quantity
  • Availability
  • Add to Cart