ENA321761900 Enamine

3-bromo-1-(2-chlorophenyl)pyrrolidin-2-one

  • MDL number MFCD18483224
  • Cas Number : 1311313-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73846
Availability details for
Quantity
  • Availability
  • Add to Cart