ENA321761901 Enamine

2-(1-aminopropyl)phenol hydrochloride

  • MDL number MFCD18483225
  • Cas Number : 1311314-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73847
Availability details for
Quantity
  • Availability
  • Add to Cart