ENA321761903 Enamine

(1-amino-4-methylpentan-2-yl)(1-phenylethyl)amine

  • MDL number MFCD18483227
  • Cas Number : 1311314-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73872
Availability details for
Quantity
  • Availability
  • Add to Cart