ENA321761904 Enamine

[3-(aminomethyl)pentan-3-yl](1-phenylethyl)amine

  • MDL number MFCD18483228
  • Cas Number : 1311315-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73874
Availability details for
Quantity
  • Availability
  • Add to Cart