ENA321761906 Enamine

2-{[(1-benzyl-1H-pyrazol-4-yl)methyl]amino}ethan-1-ol hydrochloride

  • MDL number MFCD18483230
  • Cas Number : 1311314-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73894
Availability details for
Quantity
  • Availability
  • Add to Cart