ENA321761907 Enamine

4-amino-1-(4-phenylpiperazin-1-yl)butan-1-one dihydrochloride

  • MDL number MFCD18483231
  • Cas Number : 1311316-75-2
  • Molecular Weight : 320.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73910
Availability details for
Quantity
  • Availability
  • Add to Cart