ENA321761909 Enamine

2-(2,4-dioxo-1,3-diazinan-1-yl)acetic acid

  • MDL number MFCD18483233
  • Cas Number : 1311314-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73924
Availability details for
Quantity
  • Availability
  • Add to Cart