ENA321761910 Enamine

1-(2,3-dihydro-1,4-benzodioxine-6-sulfonyl)azetidine-2-carboxylic acid

  • MDL number MFCD18483234
  • Cas Number : 1311313-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73932

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73932
Availability details for
Quantity
  • Availability
  • Add to Cart