ENA321761911 Enamine

4-ethenylpiperidine hydrochloride

  • MDL number MFCD18483236
  • Cas Number : 1311316-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73939
Availability details for
Quantity
  • Availability
  • Add to Cart