ENA321761917 Enamine

(5-methyl-1,2-oxazol-3-yl)methanesulfonyl fluoride

  • MDL number MFCD18483245
  • Cas Number : 1311317-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73995
Availability details for
Quantity
  • Availability
  • Add to Cart