ENA321761919 Enamine

4-(pyrrolidine-1-carbonyl)-1H-pyrazole hydrochloride

  • MDL number MFCD18483247
  • Cas Number : 1311316-59-2
  • Molecular Weight : 201.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73998
Availability details for
Quantity
  • Availability
  • Add to Cart