ENA321761920 Enamine

methyl 5-(chlorosulfonyl)-3-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD18483249
  • Cas Number : 1346270-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74004
Availability details for
Quantity
  • Availability
  • Add to Cart