ENA321761922 Enamine

8$l^{6}-thia-2,3,5,7-tetraazatricyclo[7.4.0.0^{2,6}]trideca-1(9),3,5,10,12-pentaene-8,8-dione

  • MDL number MFCD18917243
  • Cas Number : 1311314-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284...286
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74018

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74018
Availability details for
Quantity
  • Availability
  • Add to Cart