ENA321761935 Enamine

(2,4-dimethoxyphenyl)(4-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD18785459
  • Cas Number : 1071592-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74113
Availability details for
Quantity
  • Availability
  • Add to Cart