ENA321761937 Enamine

methyl 4-[3-(chloromethyl)-1,2,4-oxadiazol-5-yl]butanoate

  • MDL number MFCD18483255
  • Cas Number : 1311314-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74138
Availability details for
Quantity
  • Availability
  • Add to Cart