ENA321761938 Enamine

N-(benzenesulfonyl)-2,2,2-trifluoroethanecarbonimidoyl chloride

  • MDL number MFCD18483256
  • Cas Number : 642461-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74139
Availability details for
Quantity
  • Availability
  • Add to Cart