ENA321761939 Enamine

2-(3-methanesulfonyl-4-methoxyphenyl)acetonitrile

  • MDL number MFCD18483257
  • Cas Number : 1311315-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74145
Availability details for
Quantity
  • Availability
  • Add to Cart