ENA321761943 Enamine

2-bromo-1-(1,3-dimethyl-1H-pyrazol-4-yl)ethan-1-one hydrobromide

  • MDL number MFCD18483263
  • Cas Number : 1311317-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74165
Availability details for
Quantity
  • Availability
  • Add to Cart