ENA321761959 Enamine

2-(3-chlorophenyl)-2-[(2-methylpropyl)sulfanyl]ethan-1-amine hydrochloride

  • MDL number MFCD18483277
  • Cas Number : 1311314-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-74273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74273
Availability details for
Quantity
  • Availability
  • Add to Cart