ENA321761969 Enamine

2,2,2-trifluoro-1-(1-methyl-1H-pyrazol-4-yl)ethan-1-one

  • MDL number MFCD10699045
  • Cas Number : 1174886-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56201
Availability details for
Quantity
  • Availability
  • Add to Cart