ENA321761972 Enamine

[(4-chlorophenyl)methyl](ethyl)amine

  • MDL number MFCD00045212
  • Cas Number : 69957-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32133
Availability details for
Quantity
  • Availability
  • Add to Cart