ENA321761975 Enamine

(2-methoxyethyl)({1-[3-(trifluoromethyl)phenyl]ethyl})amine

  • MDL number MFCD11139382
  • Cas Number : 1019598-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32478
Availability details for
Quantity
  • Availability
  • Add to Cart